missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water (0.091 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water (0.091 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 86°C to 88°C |
Quantity | 250 mg |
UN Number | UN3261 |
Beilstein | 1285265 |
Sensitivity | Moisture sensitive |
Solubility Information | Sparingly soluble in water (0.091g/L at 25°C). |
Formula Weight | 232.71 |
Percent Purity | 97% |
Chemical Name or Material | Benzo[b]thiophene-3-sulfonyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'