Learn More
Benzoic anhydride, 98%, Thermo Scientific Chemicals
Description
Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.
Solubility
Soluble in alcohol, ether, chloroform, acetone, ethyl acetate, benzene, toluene, glacial acetic acid, acetic anhydride and ethanol. Slightly soluble in benzene and acetone. Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
Specifications
Melting Point | 40°C to 44°C |
Density | 1.199 |
Boiling Point | 360°C |
Flash Point | >110°C (230°F) |
Assay Percent Range | ≥98% |
Quantity | 50 g |
Beilstein | 516726 |
Sensitivity | Moisture sensitive |
Merck Index | 14,1092 |
Solubility Information | Soluble in alcohol,ether,chloroform,acetone,ethyl acetate,benzene,toluene,glacial acetic acid,acetic anhydride and ethanol. Slightly soluble in benzene and acetone. Insoluble in water. |
Show More |
RUO – Research Use Only