missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-6-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-4258757
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3622-35-3
C8H5NO2S
179.19
MFCD00111651
DMPZJACLHDWUFS-UHFFFAOYSA-N
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
601670
1,3-benzothiazole-6-carboxylic acid
OC(=O)C1=CC=C2N=CSC2=C1
This item is not returnable. View return policy
3622-35-3
C8H5NO2S
179.19
MFCD00111651
DMPZJACLHDWUFS-UHFFFAOYSA-N
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
601670
1,3-benzothiazole-6-carboxylic acid
OC(=O)C1=CC=C2N=CSC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 241°C to 243°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 179.2
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Benzothiazole-6-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Benzothiazole-6-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'