missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Chloride, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072863
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
98-88-4
C7H5ClO
653
This item is not returnable. View return policy
98-88-4
C7H5ClO
653
Melting Point -1°C
Color Colorless
pH 2
Boiling Point 198°C
Packaging Plastic Coated Glass bottle
Quantity 250 mL
Vapor Density 4.88
Formula Weight 140.57g/mol
Vapor Pressure 0.5 hPa at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'