missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, Multi-Compendial, N.F., J.T. Baker™
Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Specifications
Flash Point | 93.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 4 L |
Specific Gravity | 1L = 1.05kg |
Formula Weight | 108.14 |
Grade | Multi-Compendial/NF |
Chemical Name or Material | Benzyl Alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'