missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol NF, Macron Fine Chemicals™
Specifications
Specifications
| Packaging | Glass |
| Quantity | 4 L |
| Formula Weight | 108.14 |
| Grade | NF |
| Chemical Name or Material | Benzyl Alcohol |
Safety and Handling
Product Identifier
- Benzyl alcohol
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Benzyl alcohol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?