missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cyanide, 99+%, Thermo Scientific Chemicals

Catalog Number p-7214337
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
140-29-4
117.15
This item is not returnable. View return policy
140-29-4
117.15

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -24.0°C
Density 1.0100 g/mL
Boiling Point 233.0°C to 234.0°C
Flash Point 102°C
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H7N
Linear Formula C6H5CH2CN
Refractive Index 1.5220 to 1.5240
Quantity 500 mL
Beilstein 09, 441
Merck Index 15, 1134
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 117.15
Chemical Name or Material Benzyl cyanide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'