Learn More
Emma Safety Footwear Bergamo Safety Shoes
Comfortable foot-hugging fit provides a naturally light and airy foot feel. Virtually penetration proof, water resistant uppers. The 100% nitrile rubber outsole repels liquids, harmful or otherwise and the ingeniously designed sole contours provide extra grip on almost all surfaces.
Brand:  Emma Safety Footwear 107092-40
Additional Details : Weight : 1.40600kg
 View more versions of this product
Product Code. 15908171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • O3-Work shoe without steel toecap with sole plate
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • Metal-free
  • Chrome-free lining
  • ESD certified
  • Sanitized
Specifications
Specifications
EN ISO 20347:2012
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
40
Laces
Bergamo
Leather