missing translation for 'onlineSavingsMsg'
Learn More

BES sodium salt, 99%, ACROS Organics™

Brand:  Acros Organics 439035000

Code : 29

Additional Details : CAS Number : 66992-27-6 Weight : 0.55000kg

Quantity 500g

 View more versions of this product

Product Code. 11935291

  • 612.00 € / 500g

Estimated Shipment: 07-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Biological buffer
Specifications

Chemical Identifiers

66992-27-6
235.24
bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
C6H14NO5S·Na
CFQLQLSIZOWFNV-UHFFFAOYSA-M
23671640
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifications

BES sodium salt
98.5
99%
bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
235.24
235.24
99%
Authentic
155.6°C to 156.5°C
66992-27-6
100.0
C6H14NO5S·Na
CFQLQLSIZOWFNV-UHFFFAOYSA-M
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
23671640
Powder or Crystalline Powder
30.18 to 31.09% C, 5.85 to 6.04% N
White
500g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BES sodium salt

SDS Documents
Certificates