missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES sodium salt, 99%

Catalog Number p-4258710
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
66992-27-6
C6H14NO5S·Na
235.24
CFQLQLSIZOWFNV-UHFFFAOYSA-M
bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
23671640
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
66992-27-6
C6H14NO5S·Na
235.24
CFQLQLSIZOWFNV-UHFFFAOYSA-M
bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
23671640
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Melting Point 155.6°C to 156.5°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder or Powder
Quantity 25 g
Assay Percent Range 99%
Formula Weight 235.24
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material BES sodium salt
Show More Show Less
Product Identifier
  • BES sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'