missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals beta-Casomorphin, human, 95+%
Click to view available options
Quantity:
5mg
25mg
Description
An opioid peptide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Purity | ≥95% |
Color | White |
Content And Storage | Store at -20°C |
Format | Powder |
Molecular Weight (g/mol) | 864.01 |
Product Type | beta-Casomorphin |
Quantity | 5mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'