missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-D™ Sterile Disposable Overboots

Catalog Number p-9023019
Click to view available options
Description:
Standard Height
This item is not returnable. View return policy
This item is not returnable. View return policy

Single-use overboots constructed of CleanTough™ fabric are suitable for ISO 4 (Class 10) controlled environments.

  • Sole laminated with Safe-Step™ material
  • Elasticated top with secure ties
  • Sterilized by gamma irradiation
  • Low linting and anti-static property fabric
  • ISO Class 4 and EU GMP Grade A compatible
  • Universal size
  • Applications: Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities, Compounding, Manufacturing, Industrial
Size Universal
Material Spunbond Polypropylene, Poly Film
Color White
Disposable Single-use
Sterility Sterile
Closure Type Integral Ties
Sole Type Slip-resistant
Description Standard Height
Construction Single Needle Stitching
Seam Type Bound
For Use With (Application) Cleanroom, Medical Device Manufacturing, Biotechnology, Pharmaceutical
Certifications/Compliance ASTM F739, PPE Category III, ISO 11137-1
Certifications/Compliance (Europe) CE 0598, EN ISO 13688:2013, EN 1149-5, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325,
Packaging Quantity One pair per sealed inner PE bag; 15 inner bags per sealed outer PE bag; five outer bags per lined carton (75 pairs)
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'