Learn More
Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand: Corning™ 356537
Code : NEW
Additional Details : Weight : 4.07000kg
Description
Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.Specifications
8.45 oz. | |
250 mL | |
Canted | |
Rectangular | |
Sterile | |
75 cm2 |
Phenolic | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
50/cs. | |
Poly-D-Lysine | |
Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.