Learn More
IBA Lifesciences Biotin Blocking Buffer
Biotin Blocking Buffer is optimized for efficient blocking of biotinylated proteins in western blot analyses
Brand: IBA Lifesciences 2-0501-002
Code : LE
Additional Details : Weight : 0.10000kg
Description
Depending on the host organism, protein extracts may contain small amounts of biotinylated proteins such as the biotin carboxyl carrier protein (BCCP, 22 kDa) of E. coli.Biotin Blocking Buffer is recommended for blocking such biotinylated proteins on western blots prior to detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins with Strep-Tactin™ or Strep-Tactin™XT conjugates to prevent the detection of false positive bands. The buffer contains the tetrameric biotin binding protein avidin, which selectively binds biotinylated proteins. Please note that usage of BioLock is not necessary when using StrepMAB-Classic, since this antibody does not bind to biotinylated proteins.Specifications
2 mL |
For Research Use Only.