missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%, Thermo Scientific Chemicals

Catalog Number p-3759095
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-23-1
C22H42O4
370.57
MFCD00009496
SAOKZLXYCUGLFA-UHFFFAOYSA-N
bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
7641
CHEBI:34675
bis(2-ethylhexyl) hexanedioate
CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-23-1
C22H42O4
370.57
MFCD00009496
SAOKZLXYCUGLFA-UHFFFAOYSA-N
bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
7641
CHEBI:34675
bis(2-ethylhexyl) hexanedioate
CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Density 0.9240g/mL
Boiling Point 210.0°C to 218.0°C (5.0 mmHg)
Flash Point 196°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480
Quantity 250 g
Specific Gravity 0.924
Solubility Information Solubility in water: insouble. Other solubilities: soluble in aceton,ether,ethanol
Viscosity 13-15 mPa.s (20°C)
Formula Weight 370.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) adipate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'