missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 443100010

Code : NEW

Additional Details : CAS Number : 111-96-6 Weight : 0.98700kg Transport : UN number : 3271 Chem class : 3 Pack group : III


Quantity 1L

 View more versions of this product

Product Code. 11954701

  • 197.00 € / 1 L

In stock, ready to ship: 8

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

111-96-6
134.18
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
CHEBI:46784
COCCOCCOC
C6H14O3
SBZXBUIDTXKZTM-UHFFFAOYSA-N
8150
1-methoxy-2-(2-methoxyethoxy)ethane

Specifications

Bis(2-methoxyethyl) ether
51°C
1.4070 to 1.4090
2 mPa.s (20°C)
162.0°C
1L
99% min. (GC)
(CH3OCH2CH2)2O
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
8150
134.18
Extra Dry over Molecular Sieve
0.9370g/mL
Authentic
0.937
0.005% max.
-64.0°C
111-96-6
C6H14O3
15,3184
Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, hydrocarbon solvents
COCCOCCOC
134.18
CHEBI:46784
99+%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methoxyethyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H226-Flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS Documents
Certificates