missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.9%, extra pure, ACROS Organics™

Brand:  Acros Organics 208070025

Code : 29

Additional Details : CAS Number : 1304-76-3 Weight : 2.55000kg

Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11954161

  • 283.00 € / 2.50kg

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1304-76-3
Bi2O3
465.95
Bismuth trioxide

Specifications

Bismuth(III) oxide
99.9%
Bi2O3
15,1273
Solubility in water: insoluble. Other solubilities: soluble in acids
465.95
99.9%
10ppm max.
0.2% max.
50ppm max.
8.9
1890°C
825.0°C
1304-76-3
Bi2O3
01,59
Bismuth trioxide
465.95
Powder
Extra Pure
8.9000g/mL
Plastic bottle
Solubility in water: insoluble. Other solubilities: soluble in acids
10ppm max.
Yellow
2.5kg
Videos
SDS Documents
Certificates