missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) oxyiodide, 98%, pure, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Specifications
Quantity | 50 g |
Assay Percent Range | 97.5% min |
Linear Formula | IBi=O |
Merck Index | 15,127 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 351.88 |
Percent Purity | 98% |
Grade | Pure |
Chemical Name or Material | Bismuth(III) oxyiodide |
Safety and Handling
Product Identifier
- Bismuth(III) oxyiodide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'