missing translation for 'onlineSavingsMsg'
Learn More

Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific™

691.00 €

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474125
SDS View Documents
Thermo Scientific Alfa Aesar
047229.KF
1 pc.
691.00 €
Pack of 1
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]

Specifications

7440-69-9
≈7.5g/cm
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
99.9999%
(metals basis)
Odorless
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
208.98
CHEBI:33301
Puratronic™
Bismuth rod, 10mm (0.39 in.) dia. x 145mm (5.71 in.) long
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-177-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.