Learn More
Kautex™ Black Polypropylene Screw Closures
0.65 € - 109.00 €
Specifications
Product Type | Screw Cap |
---|---|
Color | Black |
Material | Polypropylene |
Liner Material | Polyethylene Foam Insert |
Product Code | Brand | Diameter (Metric) | For Use With (Equipment) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | For Use With (Equipment) | Includes | Price | Quantity & Availability | |||
15981471
|
Kautex™
2000971152 |
32 mm | 100 mL Bottle | w/out Tamper Evident Ring |
51.79 €
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
15921411
|
Kautex™
2000070750 |
32 mm | 50 mL Bottle | w/out Tamper Evident Ring |
30.40 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
15981451
|
Kautex™
2000084479 |
40 mm | 100 mL Bottle | w/Tamper Evident Ring |
49.35 €
Pack of 100 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
15931411
|
Kautex™
2000070751 |
40 mm | 100 mL Bottle | w/out Tamper Evident Ring |
38.16 €
Pack of 100 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||
15991471
|
Kautex™
2000971153 |
45 mm | 250/350 mL Bottle | w/out Tamper Evident Ring |
61.93 €
Pack of 100 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||
15941411
|
Kautex™
2000070752 |
50 mm | 200/300 mL Bottle | w/out Tamper Evident Ring |
43.26 €
Pack of 100 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||
15901481
|
Kautex™
2000971898 |
54 mm | 500/750/1000 mL Bottle | w/out Tamper Evident Ring |
81.15 €
Pack of 100 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||
15951411
|
Kautex™
2000070753 |
65 mm | 500 mL Bottle | w/out Tamper Evident Ring |
76.61 €
Pack of 100 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||
17888167
|
Kautex™
2000084481 |
65 mm | 500 mL Bottle | w/Tamper Evident Ring |
95.41 €
Pack of 100 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17898147
|
Kautex™
2000070755 |
80 mm | 2000 mL Bottle | w/out Tamper Evident Ring |
103.93 €
Pack of 100 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15961411
|
Kautex™
2000070754 |
80 mm | 750/1000 mL Bottle | w/out Tamper Evident Ring |
109.00 €
Pack of 100 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||
10491771
|
Kautex™
2000971155 |
80 mm | 1500/2500/4000 mL Bottle | w/out Tamper Evident Ring |
1.54 €
Each |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||
10628271
|
Kautex™
2000971152 |
32 mm | 100 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10632371
|
Kautex™
2000070751 |
40 mm | 100 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10569252
|
Kautex™
2000971898 |
54 mm | 500/750/1000 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15921421
|
Kautex™
2000071150 |
25 mm | 50 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10129570
|
Kautex™
2000070753 |
65 mm | 500 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10615331
|
Kautex™
2000070750 |
32 mm | 50 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10568822
|
Kautex™
2000971153 |
45 mm | 250/350 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10483511
|
Kautex™
2000070755 |
80 mm | 2000 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10360811
|
Kautex™
2000070752 |
50 mm | 200/300 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10697161
|
Kautex™
2000071150 |
25 mm | 50 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10087950
|
Kautex™
2000070754 |
80 mm | 750/1000 mL Bottle | w/out Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10749221
|
Kautex™
2000084479 |
40 mm | 100 mL Bottle | w/Tamper Evident Ring |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Specifications
Screw Cap | |
Black | |
Polypropylene | |
Polyethylene Foam Insert |