missing translation for 'onlineSavingsMsg'
Learn More

Manutan Blue medium duty steel shelving starter bay, 2000mm x 1000mm x 700mm

Brand:  Manutan A360341

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12882408

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Economic multipurpose shelving. All steel for durability with easy 'click-in' system.
  • Five shelves
  • Blue uprights
  • Shelf capacity UDL: 125kg
  • Dimensions (H x W x D): 2000mm x 1000mm x 700mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers