Learn More
Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels
Designed for use with various BMP™50 series label printers. Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels offer a repositionable adhesive and are intended for use in wire and cable marking, component or equipment identification, barcoding, and other general identification.
Brand:  Brady™ 143330
Code : NEW
Additional Details : Weight : 0.09070kg
 View more versions of this product
Product Code. 12846915
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Designed for use with various BMP™50 series label printers. Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels offer a repositionable adhesive and are intended for use in wire and cable marking, component or equipment identification, barcoding, and other general identification.
  • Adhesive and cloth backing offers excellent holding power, yet allow for clean removal and repositioning
  • Stick to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • All-in-one cartridge includes label supply and R6200 Series ribbon
Specifications
Specifications
RoHS, UL
Rubber, Repositionable
Semi-Gloss
Vinyl Cloth (B-498)
None
BMP™41, BMP™51, and BMP™53 printers
1
1 Ea.
-40 to 176°F
55 lb./in.
0.185 mm
6.10 m
9.53 mm
BMP41/BMP51/BMP53 Labelmaker Labels
Black, White
Cartridge
Thermal Transfer
Barcoding, Component ID, Equipment ID, General purpose ID, Wire and cable marking
1 (cont.)
2 Years
0.375 in. x 20 ft.
-40°C to +80°C
0.0073 in.
20 ft.
0.375 in.
6.1 m