missing translation for 'onlineSavingsMsg'
Learn More

BOC-O-Benzyl-L-threonine, 99%, Thermo Scientific™

Catalog Number p-4813358
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15260-10-3
C16H23NO5
309.36
MFCD00066062
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O
This item is not returnable. View return policy
15260-10-3
C16H23NO5
309.36
MFCD00066062
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Melting Point 115°C to 117°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 15.5
Formula Weight 309.37
Percent Purity 99%
Chemical Name or Material BOC-O-Benzyl-L-threonine
Product Identifier
  • BOC-O-Benzyl-L-threonine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'