missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PTFE Reducing Unions

Connect tubing or tubes with different outer diameters. PTFE Reducing Unions are vacuum suitable and feature universal chemical resistance.

Brand:  Bohlender™ D526-02

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10045020

  • 29.50 € / Each

Estimated Shipment: 27-10-2023
to see stock.
Add to basket

Description

Description

  • Straight PTFE tube fitting
  • Nuts made of glass-fibre reinforced PTFE
  • Universal chemical resistance
  • Product is only exposed to PTFE
  • Compliance

    FDA

  • Specifications

    Specifications

    Swivelling Screw Cap
    Autoclavable
    For 3.2mm (1/8 in.) O.D. tubing
    -50°C
    6mm O.D.; 4mm O.D.
    White/Black
    250°C
    FDA Conform
    PPS, PTFE
    Insertion of probes, tubes or tubing into vessels with GL necks for avoiding collision with the stirrer shaft
    1 Each
    GL 18
    5 bar
    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers