Learn More
Emma Safety Footwear Bologna XD Safety Shoes
A classically styled Brogue style Business shoe. Polished black full grain leather uppers and a virtually impenetrable nitrile rubber sole allows the wearer to stroll from the office to the factory and back again, with no fuss or shoe changing.
Brand: Emma Safety Footwear 107000-44
Additional Details : Weight : 1.70600kg
Description
- S3-Safety shoe with sole plate for wet work
- Nitrile rubber heat resistant outsole
- Full grain leather construction
- ESD certified
- Chrome-free lining
- Sanitized
Specifications
Bologna XD | |
Leather | |
44 |