missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21553

Additional Details : Weight : 0.39600kg

 View more versions of this product

Product Code. 10225551

  • 110.00 € / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

100mm (OD); 95mm (ID)
22mm
100 mm
100mm
Buchner filter funnel
500 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers