missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21254

Additional Details : Weight : 0.27000kg

 View more versions of this product

Product Code. 10676573

  • 98.30 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

85mm (OD); 80mm (ID)
18mm
65 mm
85mm
Buchner filter funnel
250 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers