missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21753

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10032161

  • 31.60 € / Each

Estimated Shipment: 02-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

52mm (OD); 48mm (ID)
10mm
50 mm
52mm
Buchner filter funnel
75 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
3
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers