Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Tube
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Conical Filter Tubes are used for filtration of very small volumes and is highly resistant to most chemicals such as solvents, acids, and alkalis.
Brand:  Lenz Laborglasinstrumente™ 07220004
Additional Details : Weight : 0.10000kg
Product Code. 10479172
  • 45.95 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Filter tubes
  • Made of borosilicate glass
Specifications
Specifications
Filter Tube
Borosilicate Glass
4
30 mL
20 mm