Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04000049
Code : NEW
Additional Details : Weight : 0.60000kg
Description
- Without scale; with solid glass plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
250 mL | |
Borosilicate Glass | |
Polyethylene |