missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale

For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.

Brand:  Lenz Laborglasinstrumente™ 04000058

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15330885

  • 139.00 € / Pack of 2

Call for latest local availability
01 885 5854

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

Borosilicate Glass
4.0 mm bore
Polyethylene
No
2
500 mL
Videos
Documents