missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Borosilicate Glass Filter Flask
Lateral socket for vacuum tubing with outer diameter 15-18 mm
63.15 € - 198.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10325331
|
BRAND™
73719 |
500 mL |
63.15 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10335331
|
BRAND™
73724 |
1000 mL |
82.20 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10759201
|
BRAND™
73729 |
2000 mL |
198.00 €
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
Description
Specifications
Clear | |
Borosilicate Glass | |
Lateral socket |