missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug and Graduations

Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug and Graduations are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.

Brand:  Lenz Laborglasinstrumente™ 04022028

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10579571

  • 95.72 € / Pack of 2
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

  • Cylindrical shape with straight discharge tube
  • With NS socket and PE stopper
  • With solid glass plug
  • With Screwthreade retaining device
  • With scale
Specifications

Specifications

50 mL
Yes
2mL
0.10000kg
Borosilicate Glass
Solid Glass plug and screw thread retaining device
29/32 NS Socket
Separating funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers