Learn More
Brady™ ToughStripe™ Floor Marking Tape
ToughStripe™ Floor Marking Tape (3.5m/100 ft. in length) is a long-lasting, heavy-duty solution for marking floors, aisles, work cells and other designated areas.
Brand:  Brady™ 104349
Code : NEW
Additional Details : Weight : 0.93670kg
 View more versions of this product
Product Code. 11781685
  • 122.00 € / 1 roll
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Gloss
Polyester
Used to mark aisles, passageways, storage locations and point out safety equipment and exits
1 Roll
-18°C to +54°C
0.201 mm
100 ft.
3 in.
ToughStripe FLOOR TAPES
Black, White
Tape
Non-sterile
115 Gardner Units
0°F to 130°F
0.008 in.
100 ft. x 3 in. (30.48 m x 7.62 cm)
30.48 m
7.62 cm