Learn More
Thermo Scientific Chemicals Bradykinin (1-7)
For cellular and molecular biology applications
64.20 € - 192.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427859
|
Thermo Scientific Chemicals
J66481.#0 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15437859
|
Thermo Scientific Chemicals
J66481.MB |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Bradykinin Fragment 1-7 is used in research on the specificity and kinetics of kininase(s). Bradykinin dilates blood vessels via the release of prostacyclin, nitric oxide, and Endothelium Derived Hyperpolarizing Factor. Bradykinin is a physiologically and pharmacologically active peptide of the kinin group of proteins, consisting of nine amino acids.