Learn More
BRAND™ Transferpettor™ Replacement Seals
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
29.75 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10567951
|
BRAND™
702870 |
1000 to 5000 μL |
29.75 €
Pack of 10 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
10767981
|
BRAND™
702872 |
2000 to 10,000 μL |
29.75 €
Pack of 10 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
10683561
|
BRAND™
702866 |
200 to 1000 μL |
29.75 €
Pack of 10 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
10673561
|
BRAND™
702864 |
100 to 500 μL |
29.75 €
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Aids in achieving precision and accuracy
- For the most demanding pipetting operations
- Transferpettors operates on the positive displacement principle, with piston in direct contact with the aspirated liquid
- Provided with DE-M marking
- Supplied in packs of 10
Specifications
DE-M | |
HDPE | |
10/Pk. |