Learn More
Hirschmann™ Bright Lined Counting Chambers
Includes 2 haemocytometer glass covers.
Brand:  Hirschmann™ 8100210
Additional Details : Weight : 0.03600kg
 View more versions of this product
Product Code. 10528961
  • 177.00 € / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Double net ruling
  • Bright lined
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Specifications
Counting Chamber (Fuch-Rosenthal)
0.2 mm
CE pursuant to IVD 98/79 EC
2 haemocytometer glass covers