missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific Chemicals

Catalog Number p-3770855
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
42014-51-7
C6H6BrNO4
236.02
MFCD00058571
NKUZQMZWTZAPSN-UHFFFAOYSA-N
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
3565210
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
C1CC(=O)N(C1=O)OC(=O)CBr
This item is not returnable. View return policy
42014-51-7
C6H6BrNO4
236.02
MFCD00058571
NKUZQMZWTZAPSN-UHFFFAOYSA-N
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
3565210
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
C1CC(=O)N(C1=O)OC(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Elemental Analysis 88% min.
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 236.02
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Bromoacetic acid N-hydroxysuccinimide ester
compliance-icons
Product Identifier
  • Bromoacetic acid N-hydroxysuccinimide ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'