missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-72126
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
598-21-0
C2H2Br2O
201.85
MFCD00000115
LSTRKXWIZZZYAS-UHFFFAOYSA-N
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
61147
2-bromoacetyl bromide
C(C(=O)Br)Br
598-21-0
C2H2Br2O
201.85
MFCD00000115
LSTRKXWIZZZYAS-UHFFFAOYSA-N
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
61147
2-bromoacetyl bromide
C(C(=O)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 2.3100g/mL
Boiling Point 147°C to 150°C
Flash Point >105°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COBr
Refractive Index 1.5450 to 1.547
Quantity 100 mL
Beilstein 02, 215
Specific Gravity 2.31
Solubility Information Solubility in water: reacts. Other solubilities: solulbe in acetone,carbontetrachloride
Formula Weight 201.85
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bromoacetyl bromide, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Bromoacetyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'