missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, sodium salt, pure, water soluble, indicator

Product Code. 10225530 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10225530

Brand: Thermo Scientific Chemicals 151350250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >300.0°C
CAS 62625-28-9
Molecular Formula C19H9Br4NaO5S
MDL Number MFCD00013793
Beilstein 19, I, 649
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.942
PubChem CID 3791690
Formula Weight 691.94
Grade Pure
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Bromophenol Blue, sodium salt, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.