missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, Pure, Indicator Grade, Fisher Chemical™

C27H28Br2O5S, CAS Number-76-59-5, 5g, Brown to Red, 624.38g/mol, 204 deg.C, 5872, Glass Jar, Solid

Brand:  Fisher Chemical B/4690/44

Code : 21

Additional Details : CAS Number : 76-59-5 Weight : 0.05000kg

Product Code. 10273370

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

C27H28Br2O5S, CAS Number-76-59-5, 5g, Brown to Red, 624.38g/mol, 204 deg.C, 5872, Glass Jar, Solid

Specifications

Specifications

204°C
C27H28Br2O5S
624.38
Pure
Brown to Red
Solid
76-59-5
5872
624.38g/mol
Glass Jar
5 g
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers