missing translation for 'onlineSavingsMsg'
Learn More

Brucine sulfate heptahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-77752
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Packaging:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
60583-39-3
C46H68N4O19S
1013.12
MFCD00150159
PPJIVFYMRNHHTJ-UMRIXEKQSA-N
brucinesulfateheptahydrate fornitrateanalysis
126969999
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate
O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
60583-39-3
C46H68N4O19S
1013.12
MFCD00150159
PPJIVFYMRNHHTJ-UMRIXEKQSA-N
brucinesulfateheptahydrate fornitrateanalysis
126969999
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate
O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Ignition Residue 0.1% max.
Loss on Drying 13.0% max. (1 g, 105°C, 6 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 10 g
Sensitivity (to nitrate) Passes Test
Merck Index 15, 1464
Formula Weight 1013.13
Clarity Passes test
Percent Purity 98+%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Brucine sulfate heptahydrate
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Brucine sulfate heptahydrate
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Mention de danger
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'