Learn More
Brucine sulfate heptahydrate, 98+%, ACS reagent, ACROS Organics™
Brand: Acros Organics 403290100
Code : 29
Additional Details : CAS Number : 4845-99-2 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : I
Packaging | Glass bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
60583-39-3 | |
1013.13 | |
GKMNADXKDGDKNA-RVZKJRTISA-N | |
126969999 | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O.O.O.O.O.O.OS(=O)(=O)O |
H2SO4·7H2O | |
MFCD00150159 | |
brucinesulfateheptahydrate fornitrateanalysis | |
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate |
Specifications
Passes Test | |
Passes test | |
Authentic | |
Glass bottle | |
(to nitrate) Passes Test | |
10g | |
98+% | |
MFCD00150159 | |
brucinesulfateheptahydrate fornitrateanalysis | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O.O.O.O.O.O.OS(=O)(=O)O | |
1013.13 | |
1013.13 | |
98+% |
Brucine sulfate heptahydrate | |
0.1% max. | |
13.0% max. (1 g, 105°C, 6 hrs) | |
0.1% max | |
White | |
60583-39-3 | |
H2SO4·7H2O | |
15, 1464 | |
GKMNADXKDGDKNA-RVZKJRTISA-N | |
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate | |
126969999 | |
Crystalline Powder | |
ACS Reagent |
Safety and Handling
- Brucine sulfate heptahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H330-Fatal if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.