Learn More
BUCHI Cold Trap Outer Part
Outer part for Rotavapor™ systems
Brand: BUCHI 025614
Code : 61
Additional Details : Weight : 0.86000kg
Description
For evaporation methods with the Rotavapor
- Includes: tube clip GL-14 complete
- For use with condenser: CR
- Coating: P+G
Specifications
Si-Cold Trap | |
Cold trap | |
Tube connection GL-14 complete | |
Plastic and Glass |