missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Evaporating Flask and Accessories

Flasks made of borosilicate 3.3 glass which is resistant to chemicals and temperature changes of 180°C

Brand:  BUCHI 001758

Code : A0

Additional Details : Weight : 1.07000kg

Product Code. 10047101

  • 200.00 € / 1 L

Estimated Shipment: 16-06-2022
to see stock.
Add to basket

Description

Description

  • Flask ring made of non-sliding silicone
Specifications

Specifications

Silicone Oil
Evaporation system
1000 mL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers