missing translation for 'onlineSavingsMsg'
Learn More

Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Catalog Number p-8015114
31.38 EUR valid until 2024-12-31
Use promo code "21980" to get your promotional price.


31.38 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.


45.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container

Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 7, MDL Number: 81857, Physical Form: Liquid

Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
  • Standard and color-coded buffer solutions
  • Manufactured to a tolerance of ±0.02 pH units
  • Accurately standardized and labelled with lot-specific pH at 20°C
  • All buffers are traceable to NIST and BS 1647 standard
  • Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
  • Standard buffers are available over the range pH2 to pH10
  • Color-coded buffer solutions are available over the range pH4, pH7, and pH
TRUSTED_SUSTAINABILITY
Color Yellow
pH 7
Physical Form Liquid
Quantity 500 mL
MDL Number 81857
Packaging HDPE plastic bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'