Learn More
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 7, MDL Number: 81857, Physical Form: Liquid
Brand: Fisher Chemical J/2855/08
12.26 EUR valid until 2021-04-30
Use promo code "14803" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Packaging | HDPE plastic bottle |
---|---|
Quantity | 500mL |
Description
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 7, MDL Number: 81857, Physical Form: Liquid
Ready to use for pH measurement.Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH
Chemical Identifiers
81857 |
Specifications
81857 | |
HDPE plastic bottle | |
Yellow |
Liquid | |
7 | |
500mL |