Learn More
Buffer, Precision Reference Standard, pH 10.000 ± 0.005 at 25°C, Ricca Chemical
Standardized against NIST-certified pH references. High-Accuracy pH Buffer solutions are for use with meters with 0.001 resolution.
Brand: Cole Parmer 05942-69
Code : A0
Additional Details : CAS Number : 1310-73-2 Weight : 0.80000kg
Description
- High-accuracy liquid 10.000 pH buffer
- Sold in individual bottles of 500 mL, or as a case of twelve.
- Store away from direct sunlight
Specifications
Buffer, Precision Reference Standard | |
0°C | |
100°C | |
10 | |
500 mL | |
Miscible |
pH 10.000 ± 0.005 at 25 deg C (Color Coded Blue) | |
497-19-8,7732-18-5,1310-73-2 | |
Blue | |
Liquid | |
Clear PET-G Bottle |