missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 10, Reagecon™
BIOPHARMA

Catalog Number 11474559
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy
pH 10
Quantity 1 L
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 10.00 ± 0.01 @ 25°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'