Learn More
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
27.70 € - 81.40 €
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410171
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
038714.AP
500 mL
27.70 €
500mL
View alternative products
Save Up to 
»
In Stock
to see stock.
11398987
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
038714.K2
1 L
44.00 €
1 L
View alternative products
Save Up to 
»
In Stock
to see stock.
11308997
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
038714.K7
4 L
81.40 €
4 L
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.